accessibility menu, dialog, popup

UserName

Viewing profile as user:
HN

CAS RN 532-18-3

IUPAC Name: N-(naphthalen-2-yl)naphthalen-2-amine
Synonyms: b,b'-dinaphthylamine
Molecular Weight (g/mol): 269.35
Molecular Formula: C20H15N
InChi Key: SBMXAWJSNIAHFR-UHFFFAOYSA-N
SMILES: N(C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)