accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 532-34-3

IUPAC Name: butyl 2,2-dimethyl-4-oxo-3,4-dihydro-2H-pyran-6-carboxylate
Synonyms: indalone
Molecular Weight (g/mol): 226.27
Molecular Formula: C12H18O4
InChi Key: OKIJSNGRQAOIGZ-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC(=O)CC(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results