accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5321-49-3

IUPAC Name: 1-(2-phenylethyl)piperazine
Synonyms: 1-(2-phenylethyl)piperazine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: LKUAPSRIYZLAAO-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results