accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOCl

CAS RN 53220-41-0

IUPAC Name: 4-(4-chlorobenzoyl)piperidine
Synonyms: 4-(4-chlorobenzoyl)piperidine
Molecular Weight (g/mol): 223.7
Molecular Formula: C12H14ClNO
InChi Key: IYGWDOXHCPQXKN-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results