accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5323-87-5

IUPAC Name: 3-ethoxycyclohex-2-en-1-one
Synonyms: 3-ethoxycyclohex-2-en-1-one
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: JWCFJPLIRVYENQ-UHFFFAOYSA-N
SMILES: CCOC1=CC(=O)CCC1
Boiling Point 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (4)
  • (3)
  • (2)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)