accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5325-20-2

IUPAC Name: 3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: GTFMIJNXNMDHAB-UHFFFAOYSA-N
SMILES: O=C1CSC2=CC=CC=C2N1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)