CAS RN 5325-88-2
IUPAC Name:
naphthalene-1,5-dithiol
Synonyms:
naphthalene-1,5-dithiol
Molecular Weight (g/mol):
192.29
Molecular Formula:
C10H8S2
InChi Key:
AAPAGLBSROJFGM-UHFFFAOYSA-N
SMILES:
SC1=CC=CC2=C(S)C=CC=C12
1
–
1
of
1
results
1
1,5-Dimercaptonaphthalene 98.0+%, TCI America™
CAS: 5325-88-2 Molecular Formula: C10H8S2 Molecular Weight (g/mol): 192.294 MDL Number: MFCD00068909 InChI Key: AAPAGLBSROJFGM-UHFFFAOYSA-N Synonym: 1,5-Naphthalenedithiol PubChem CID: 21397 IUPAC Name: naphthalene-1,5-dithiol SMILES: C1=CC2=C(C=CC=C2S)C(=C1)S