accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSNH2N

CAS RN 53268-33-0

IUPAC Name: 6-aminopyridine-3-carbothioamide
Synonyms: 6-aminopyridine-3-carbothioamide
Molecular Weight (g/mol): 153.2
Molecular Formula: C6H7N3S
InChi Key: LCKBJNNMWMUYAB-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(N)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results