accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrBr

CAS RN 533-98-2

IUPAC Name: 1,2-dibromobutane
Synonyms: 1,2-dibromobutane
Molecular Weight (g/mol): 215.92
Molecular Formula: C4H8Br2
InChi Key: CZWSZZHGSNZRMW-UHFFFAOYNA-N
SMILES: CCC(Br)CBr
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)