accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5330-38-1

IUPAC Name: 4-chloro-2-(hydroxymethyl)phenol
Synonyms: 4-chloro-2-(hydroxymethyl)phenol
Molecular Weight (g/mol): 158.58
Molecular Formula: C7H7ClO2
InChi Key: AFPSETMCMFRVPG-UHFFFAOYSA-N
SMILES: OCC1=CC(Cl)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results