accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)NHOOCH3CH3H3CHOO

CAS RN 53308-95-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]pentanoic acid|BOC-NVA-OH
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: INWOAUUPYIXDHN-ZETCQYMHSA-N
SMILES: CCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)