accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5331-37-3

IUPAC Name: 2-(butylsulfanyl)ethan-1-ol
Synonyms: 2-(butylsulfanyl)ethanol
Molecular Weight (g/mol): 134.24
Molecular Formula: C6H14OS
InChi Key: ARRJJHJQSNPJFV-UHFFFAOYSA-N
SMILES: CCCCSCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results