accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5331-43-1

IUPAC Name: (benzyloxy)carbohydrazide
Synonyms: benzyloxycarbohydrazide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: RXUBZLMIGSAPEJ-UHFFFAOYSA-N
SMILES: NNC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)