accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrN

CAS RN 5332-24-1

IUPAC Name: 3-bromoquinoline
Synonyms: 3-bromoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: ZGIKWINFUGEQEO-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C=CC=CC2=C1
Boiling Point 
  • (4)
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)