accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5333-34-6

IUPAC Name: 4-(3,4-dimethoxyphenyl)-4-oxobutanoate
Synonyms: 4-(3,4-dimethoxyphenyl)-4-oxobutanoate
Molecular Weight (g/mol): 237.23
Molecular Formula: C12H13O5
InChi Key: BUAYFJKMVBIPKA-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1OC)C(=O)CCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results