accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3

CAS RN 5333-42-6

IUPAC Name: 2-octyldodecan-1-ol
Synonyms: octyldodecanol
Molecular Weight (g/mol): 298.56
Molecular Formula: C20H42O
InChi Key: LEACJMVNYZDSKR-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC(CO)CCCCCCCC
Boiling Point 
  • (2)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Grade 
  • (1)
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)