accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOO

CAS RN 5333-84-6

IUPAC Name: 4-methyl-1,3,3a,4,7,7a-hexahydro-2-benzofuran-1,3-dione
Synonyms: 4-methyl-3a,4,7,7a-tetrahydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: XPEKVUUBSDFMDR-UHFFFAOYNA-N
SMILES: CC1C=CCC2C1C(=O)OC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results