accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)HNSOONCH3H3COHONH2

CAS RN 53332-27-7

IUPAC Name: (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]propanoic acid; cyclohexanamine
Synonyms: (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]propanoic acid; cyclohexylamine
Molecular Weight (g/mol): 421.56
Molecular Formula: C21H31N3O4S
InChi Key: CZUBQNQWSOLSJH-PPHPATTJSA-N
SMILES: NC1CCCCC1.C[C@H](NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results