accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5335-87-5

IUPAC Name: 1-methoxy-4-[(4-methoxyphenyl)disulfanyl]benzene
Synonyms: 1-methoxy-4-[(4-methoxyphenyl)disulfanyl]benzene
Molecular Weight (g/mol): 278.38
Molecular Formula: C14H14O2S2
InChi Key: PZQGLCGLPMWYBT-UHFFFAOYSA-N
SMILES: COC1=CC=C(SSC2=CC=C(OC)C=C2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)