accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBOH3COCH3

CAS RN 5337-36-0

IUPAC Name: trihexyl borate
Synonyms: boric acid (H3BO3), trihexyl ester
Molecular Weight (g/mol): 314.32
Molecular Formula: C18H39BO3
InChi Key: KDQYHGMMZKMQAA-UHFFFAOYSA-N
SMILES: CCCCCCOB(OCCCCCC)OCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results