accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBOH3COCH3

CAS RN 5337-41-7

IUPAC Name: trioctadecyl borate
Synonyms: trioctadecyl borate
Molecular Weight (g/mol): 819.29
Molecular Formula: C54H111BO3
InChi Key: GZKLCETYSGSMRA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOB(OCCCCCCCCCCCCCCCCCC)OCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results