accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5339-01-5

IUPAC Name: 1-[3-(dimethylamino)phenyl]ethan-1-ol
Synonyms: 1-[3-(dimethylamino)phenyl]ethanol
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: XYGWUVVZDWJPBW-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC(=C1)N(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results