accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5339-85-5

NH2HO

CAS RN 5339-85-5

IUPAC Name: 2-(2-aminophenyl)ethan-1-ol
Synonyms: 2-(2-aminophenyl)ethanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ILDXSRFKXABMHH-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1CCO
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)