accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53397-65-2

IUPAC Name: tricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-2,10-diyne
Synonyms: tricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-2,10-diyne
Molecular Weight (g/mol): 200.24
Molecular Formula: C16H8
InChi Key: JFYLRMKUYCIWMX-UHFFFAOYSA-N
SMILES: C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results