accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)H3CBr

CAS RN 534-00-9

IUPAC Name: (2S)-1-bromo-2-methylbutane
Synonyms: (2S)-1-bromo-2-methylbutane
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: XKVLZBNEPALHIO-YFKPBYRVSA-N
SMILES: CC[C@H](C)CBr
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)