accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOCl

CAS RN 534-07-6

IUPAC Name: 1,3-dichloropropan-2-one
Synonyms: 1,3-dichloroacetone
Molecular Weight (g/mol): 126.96
Molecular Formula: C3H4Cl2O
InChi Key: SUNMBRGCANLOEG-UHFFFAOYSA-N
SMILES: ClCC(=O)CCl
Boiling Point 
  • (6)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (9)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (6)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (7)
  • (6)
  • (3)
  • (2)
  • (9)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)