accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 534-59-8

H3COHOHOO

CAS RN 534-59-8

IUPAC Name: 2-butylpropanedioic acid
Synonyms: butyl malonic acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: MCRZWYDXIGCFKO-UHFFFAOYSA-N
SMILES: CCCCC(C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)