accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3CH3H3C

CAS RN 5340-78-3

IUPAC Name: ethyl 3,3-dimethylbutanoate
Synonyms: ethyl 3,3-dimethylbutanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: JWMNHAMYTBAUPI-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(C)(C)C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)