accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53408-95-0

IUPAC Name: trimagnesium(2+) diphosphate
Synonyms: magnesium phosphate|trimagnesium phosphate
Molecular Weight (g/mol): 262.86
Molecular Formula: Mg3O8P2
InChi Key: GVALZJMUIHGIMD-UHFFFAOYSA-H
SMILES: [Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)