accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OH(S)CH3HO

CAS RN 5341-95-7

IUPAC Name: (2R,3S)-butane-2,3-diol
Synonyms: (R,R)-2,3-butanediol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: OWBTYPJTUOEWEK-ZXZARUISSA-N
SMILES: C[C@@H](O)[C@H](C)O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results