accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOH

CAS RN 5343-92-0

IUPAC Name: pentane-1,2-diol
Synonyms: pentane-1,2-diol pentylene glycol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: WCVRQHFDJLLWFE-UHFFFAOYNA-N
SMILES: CCCC(O)CO
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)