CAS RN 5344-49-0
CAS RN 5344-49-0
IUPAC Name:
                                            
                                                    2-chloro-6-nitrobenzoic acid
                                            
                                        
                                    
                                    
                                    
                                    
                                        Synonyms:
                                        
                                                2-chloro-6-nitrobenzoic acid
                                        
                                    
                                                Molecular Weight (g/mol):
                                                
                                                           201.56
                                                
                                            
                                            
                                                 Molecular Formula:
                                                
                                                        C7H4ClNO4
                                            
                                                
                                            
                                        InChi Key:
                                                
                                                        JYHOMEFOTKWQPN-UHFFFAOYSA-N
                                                
                                            
                                            
                                            SMILES:
                                                
                                                            OC(=O)C1=C(Cl)C=CC=C1[N+]([O-])=O
                                                
                                            
                                        
                                        1
                                        –
                                        1
                                        of
                                        1
                                        results
                                    
                                
                                        1 
                                    
                            
                                    
                                                        Thermo Scientific Chemicals 2-Chloro-6-nitrobenzoic acid, 99%
                                                
                                                
                                                
                                                    
                                                
                                                    
                                                
                                                                        CAS: 5344-49-0 Molecular Formula: C7H4ClNO4 Molecular Weight (g/mol): 201.562 MDL Number: MFCD00100425 InChI Key: JYHOMEFOTKWQPN-UHFFFAOYSA-N Synonym: 2-chloro-6-nitrobenzoic acid,2-chloro-6-nitro-benzoic acid,6-chloro-2-nitrobenzoic acid,benzoic acid, 2-chloro-6-nitro,pubchem20037,6-chloro-2-nitrobenzo,acmc-209l7b,2-chloro-6-nitrobenzoicacid,2-carboxy-3-chloronitrobenzene,2-chloro-6-nitrobenzoic acid # PubChem CID: 79287 SMILES: C1=CC(=C(C(=C1)Cl)C(=O)O)[N+](=O)[O-]
