accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5345-54-0

IUPAC Name: 3-chloro-4-methoxyaniline
Synonyms: 3-chloro-4-methoxyaniline
Molecular Weight (g/mol): 157.6
Molecular Formula: C7H8ClNO
InChi Key: XQVCBOLNTSUFGD-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results