accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5346-38-3

IUPAC Name: pyridine-2-carbothioamide
Synonyms: 2-pyridinethiocarboxamide
Molecular Weight (g/mol): 138.19
Molecular Formula: C6H6N2S
InChi Key: HYKQYVSNFPWGKQ-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)