accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5346-53-2

IUPAC Name: 5-amino-1-(2-hydroxyethyl)-1H-pyrazole-4-carbonitrile
Synonyms: 5-amino-1-(2-hydroxyethyl)pyrazole-4-carbonitrile
Molecular Weight (g/mol): 152.16
Molecular Formula: C6H8N4O
InChi Key: FOZZJAVSCUBGPM-UHFFFAOYSA-N
SMILES: NC1=C(C=NN1CCO)C#N
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results