accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3OOOCH3OH3COCH3

CAS RN 53464-72-5

IUPAC Name: hydrogen 8-(diethylamino)octyl 3,4,5-trimethoxybenzoate chloride
Synonyms: hydrogen 8-(diethylamino)octyl 3,4,5-trimethoxybenzoate chloride
Molecular Weight (g/mol): 432
Molecular Formula: C22H38ClNO5
InChi Key: KFJZVXKPPQIYCG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCCCCCCCOC(=O)C1=CC(OC)=C(OC)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results