accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOOH

CAS RN 53484-17-6

IUPAC Name: 1-methyl-1H-1,3-benzodiazole-5-carboxylic acid
Synonyms: 1-methyl-1,3-benzodiazole-5-carboxylic acid
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: KAJLCBKTWRUQOI-UHFFFAOYSA-N
SMILES: CN1C=NC2=CC(=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results