accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53484-19-8

IUPAC Name: ethyl 1-methyl-1H-1,3-benzodiazole-5-carboxylate
Synonyms: ethyl 1-methyl-1,3-benzodiazole-5-carboxylate
Molecular Weight (g/mol): 204.23
Molecular Formula: C11H12N2O2
InChi Key: MQBXKIFEKNCNJX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C2N(C)C=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results