accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClCl

CAS RN 535-15-9

IUPAC Name: ethyl 2,2-dichloroacetate
Synonyms: ethyl dichloroacetate
Molecular Weight (g/mol): 156.99
Molecular Formula: C4H6Cl2O2
InChi Key: IWYBVQLPTCMVFO-UHFFFAOYSA-N
SMILES: CCOC(=O)C(Cl)Cl
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results