accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3SSHN

CAS RN 5351-51-9

IUPAC Name: 4,5-dimethyl-2,3-dihydro-1,3-thiazole-2-thione
Synonyms: 4,5-dimethyl-3H-1,3-thiazole-2-thione
Molecular Weight (g/mol): 145.24
Molecular Formula: C5H7NS2
InChi Key: KKHBRTFQIYIHEI-UHFFFAOYSA-N
SMILES: CC1=C(C)SC(=S)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results