accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53515-17-6

FFFSH2N

CAS RN 53515-17-6

IUPAC Name: 3-(trifluoromethyl)benzene-1-carbothioamide
Synonyms: 3-(trifluoromethyl)benzenecarbothioamide
Molecular Weight (g/mol): 205.2
Molecular Formula: C8H6F3NS
InChi Key: WOJQBHSZMFRQIR-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(=CC=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results