accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(R)OO

CAS RN 53558-93-3

IUPAC Name: (2R)-5-oxooxolane-2-carboxylate
Synonyms: (2R)-5-oxooxolane-2-carboxylate
Molecular Weight (g/mol): 129.09
Molecular Formula: C5H5O4
InChi Key: QVADRSWDTZDDGR-GSVOUGTGSA-M
SMILES: [O-]C(=O)[C@H]1CCC(=O)O1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)