accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHOONH2(S)HOOO

CAS RN 53587-11-4

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Synonyms: benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate; para-toluene sulfonate
Molecular Weight (g/mol): 443.51
Molecular Formula: C23H25NO6S
InChi Key: PJGVHBLZZQDFFM-RSAXXLAASA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.N[C@@H](CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)