accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH2S

CAS RN 536-33-4

IUPAC Name: 2-ethylpyridine-4-carbothioamide
Synonyms: ethionamide etionizin trecator-SC etionizina etionizine ethylisothiamide etionamid ethina teberus thianid
Molecular Weight (g/mol): 166.24
Molecular Formula: C8H10N2S
InChi Key: AEOCXXJPGCBFJA-UHFFFAOYSA-N
SMILES: CCC1=CC(=CC=N1)C(N)=S
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results