accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 536-60-7

IUPAC Name: [4-(propan-2-yl)phenyl]methanol
Synonyms: cuminol cuminyl alcohol cuminic alcohol cumin alcohol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: OIGWAXDAPKFNCQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(CO)C=C1
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)