accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOBr

CAS RN 53631-18-8

IUPAC Name: 2-bromo-1-(3-fluorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(3-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: ITAQNNGDCNFGID-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results