CAS RN 536755-37-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Smopex™-111, metal scavenger, Thermo Scientific™
CAS: 536755-37-0 Synonym: Styryl thiol grafted polyethylene fiber