accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHHN

CAS RN 5368-28-5

IUPAC Name: 3-phenylpiperazin-2-one
Synonyms: 3-phenylpiperazin-2-one
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: WKFFHKBGGZHQAX-UHFFFAOYNA-N
SMILES: O=C1NCCNC1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results