CAS RN 5369-16-4
IUPAC Name:
3-(propan-2-yl)aniline
Synonyms:
3-isopropylaniline
Molecular Weight (g/mol):
135.21
Molecular Formula:
C9H13N
InChi Key:
XCCNRBCNYGWTQX-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC(N)=CC=C1
1
–
1
of
1
results
1
3-Isopropylaniline 98.0+%, TCI America™
CAS: 5369-16-4 Molecular Formula: C9H13N Molecular Weight (g/mol): 135.21 MDL Number: MFCD00092918 InChI Key: XCCNRBCNYGWTQX-UHFFFAOYSA-N Synonym: 3-Aminocumene, m-Cumidine PubChem CID: 458679 IUPAC Name: 3-(propan-2-yl)aniline SMILES: CC(C)C1=CC(N)=CC=C1