accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 537-00-8

IUPAC Name: dicerium(3+) hexaacetate trihydrate
Synonyms: dicerium(3+) hexaacetate trihydrate
Molecular Weight (g/mol): 688.54
Molecular Formula: C12H24Ce2O15
InChi Key: FLEYCIHWCBGBLG-UHFFFAOYSA-H
SMILES: O.O.O.[Ce+3].[Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)